Diethyl bromomalonate ≥92%

Synonyms: Diethyl 2-bromomalonate

D91209-100G D91209-25G
220123-271EA 323.08 USD
220123-271 220123-272
Diethyl bromomalonate ≥92%
Diethyl bromomalonate
Formula: C₇H₁₁BrO₄
MW: 239.07 g/mol
MDL Number: MFCD00009138
CAS Number: 685-87-0
EINECS: 211-683-1

Order Now

Specification Test Results

Assay 92%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR