meso-2,3-Dimercaptosuccinic acid

Synonyms: (R,S)-2,3-Dimercaptobutanedioic acid, meso-2,3-Dimercaptobutanedioic acid, Succimer, (R,S)-2,3-Dimercaptosuccinic acid, (2R,3S)-rel-2,3-Dimercaptosuccinic acid

D7881-25G D7881-50G D7881-5G D7881-1G
220122-723EA 535.23 USD
220122-723 220122-724 220122-725 220122-722
meso-2,3-Dimercaptosuccinic acid
meso-2,3-Dimercaptosuccinic acid
Formula: C₄H₆O₄S₂
MW: 182.22 g/mol
Melting Pt: 210…211 °C
Storage Temperature: Freezer
MDL Number: MFCD00064799
CAS Number: 304-55-2
EINECS: 206-155-2
Merck Index: 12,09034

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR