2,5-Dichloro-p-xylene ≥97%

D76404-25G D76404-100G
220122-972EA 343.38 USD
220122-972 220122-971
2,5-Dichloro-p-xylene ≥97%
2,5-Dichloro-p-xylene
Formula: C₈H₈Cl₂
MW: 175.06 g/mol
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 71 °C
Density: 1.39 g/cm³ (20 °C)
MDL Number: MFCD00000610
CAS Number: 1124-05-6
EINECS: 214-387-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR