2,5-Dichlorotoluene ≥98%

Synonyms: 1,4-Dichloro-2-methylbenzene

D75955-5G D75955-100G
220122-962EA 343.38 USD
220122-962 220122-961
2,5-Dichlorotoluene ≥98%
2,5-Dichlorotoluene
Formula: C₇H₆Cl₂
MW: 161.03 g/mol
Boiling Pt: 199 °C (1013 hPa)
Density: 1.2535 g/cm³ (20 °C)
MDL Number: MFCD00000609
CAS Number: 19398-61-9
EINECS: 243-032-2
UN: 3082
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR