2,5-Dichlorothiophene

D75807-100G D75807-25G
220122-959EA 106.72 USD
220122-959 220122-960
2,5-Dichlorothiophene
2,5-Dichlorothiophene
Formula: C₄H₂Cl₂S
MW: 153.03 g/mol
Boiling Pt: 162 °C (1013 hPa)
Melting Pt: –40 °C
Density: 1.4473 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005423
CAS Number: 3172-52-9
EINECS: 221-638-8
UN: 2929
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR