3,5-Dichloropyridine

D73804-25G D73804-100G
220122-926EA 44.48 USD
220122-926 220122-925
3,5-Dichloropyridine
3,5-Dichloropyridine
Formula: C₅H₃Cl₂N
MW: 147.99 g/mol
Boiling Pt: 178…179 °C (1013 hPa)
Melting Pt: 64…65 °C
MDL Number: MFCD00006376
CAS Number: 2457-47-8
EINECS: 219-537-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR