Dichloro(phenyl)phosphine

Danger

Synonyms: P,P-Dichlorophenylphosphine, Phenyldichlorophosphine

D71984-100G D71984-500G D71984-5G
220122-883EA 37.94 USD
220122-883 220122-884 220122-885
Dichloro(phenyl)phosphine
Dichloro(phenyl)phosphine
Formula: C₆H₅Cl₂P
MW: 178.98 g/mol
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: –51 °C
Density: 1.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000528
CAS Number: 644-97-3
EINECS: 211-425-8
UN: 2798
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR