4,5-Dichloro-o-phenylenediamine ≥97%

Synonyms: 4,5-Dichloro-1,2-phenylenediamine, 1,2-Diamino-4,5-dichlorobenzene, 4,5-Dichlorobenzene-1,2-diamine

D71607-25G D71607-5G
220121-761EA 412.46 USD
220121-761 220121-762
4,5-Dichloro-o-phenylenediamine ≥97%
4,5-Dichloro-o-phenylenediamine
Formula: C₆H₆Cl₂N₂
MW: 177.03 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 163 °C
MDL Number: MFCD00007723
CAS Number: 5348-42-5
EINECS: 226-305-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR