3,4-Diaminopyridine

Danger

Synonyms: 3,4-Pyridinediamine, Amifampridine, 3,4-DAP, Pyridine-3,4-diyldiamine, Pyridine-3,4-diamine

D7148-1G D7148-5G
220122-881EA 39.85 USD
220122-881 220122-882
3,4-Diaminopyridine
3,4-Diaminopyridine
Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 262 °C (1013 hPa)
Melting Pt: 218…219 °C
Flash Pt: 240 °C
Storage Temperature: Ambient
MDL Number: MFCD00006401
CAS Number: 54-96-6
EINECS: 200-220-9
UN: 2811
ADR: 6.1,II
Merck Index: 13,03006

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR