(2,4-Dichlorophenoxy)acetic acid ≥97%

Danger

Synonyms: 2,4-D

D70724-100G D70724-5G
220122-866EA 75.38 USD
220122-866 220122-867
(2,4-Dichlorophenoxy)acetic acid ≥97%
(2,4-Dichlorophenoxy)acetic acid
Formula: (C₆H₃Cl₂)OCH₂COOH
MW: 221.04 g/mol
Boiling Pt: 346 °C (1013 hPa)
Melting Pt: 137…139 °C
Density: 1.563 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004300
CAS Number: 94-75-7
EINECS: 202-361-1
Merck Index: 13,02825

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR