2,6-Dichlorophenol ≥99%

Warning

D70201-25G D70201-100G
220122-078EA 42 USD
220122-078 220122-077
2,6-Dichlorophenol ≥99%
2,6-Dichlorophenol
Formula: C₆H₄Cl₂O
MW: 163 g/mol
Boiling Pt: ∼219 °C (1013 hPa)
Melting Pt: 68 °C
Density: 1.653 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002176
CAS Number: 87-65-0
EINECS: 201-761-3
UN: 2020
ADR: 6.1,III
Merck Index: 13,03099

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR