Dicloran (2,6-dichloro-4-nitroaniline) ≥96%

Danger

Synonyms: 2,6-Dichloro-4-nitroaniline

D67820-100G D67820-5G
220122-809EA 68.6 USD
220122-809 220122-810
Dicloran (2,6-dichloro-4-nitroaniline) ≥96%
Dicloran (2,6-dichloro-4-nitroaniline)
Formula: C₆H₄Cl₂N₂O₂
MW: 207.02 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: 186…189 °C
Storage Temperature: Ambient
MDL Number: MFCD00007677
CAS Number: 99-30-9
EINECS: 202-746-4
UN: 2237
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR