Dichlone

Warning

Synonyms: 2,3-Dichloro-1,4-naphthoquinone

D67200-100G D67200-5G
220122-802EA 65.54 USD
220122-802 220122-803
Dichlone
Dichlone
Formula: C₁₀H₄Cl₂O₂
MW: 227.05 g/mol
Boiling Pt: 304 °C (1013 hPa)
Melting Pt: 193 °C
Density: 1.44 g/cm³ (16 °C)
MDL Number: MFCD00001677
CAS Number: 117-80-6
EINECS: 204-210-5
UN: 2811
ADR: 6.1,III
Merck Index: 13,03071

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR