2,4-Dichloro-5-methylpyrimidine

Danger

D66182-1G D66182-5G
220122-788EA 37.05 USD
220122-788 220122-789
2,4-Dichloro-5-methylpyrimidine
2,4-Dichloro-5-methylpyrimidine
Formula: C₅H₄Cl₂N₂
MW: 163.01 g/mol
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 26 °C
Density: 1.39 g/cm³ (25 °C)
MDL Number: MFCD00023197
CAS Number: 1780-31-0
EINECS: 217-227-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR