Diethylenetriaminepentaacetic acid ≥99%

Warning

Synonyms: N-Carboxymethyliminobis(ethylenenitrilo)tetra(acetic acid), Kestranal® Aza Pa, DTPA, Titriplex® V, Idranal V, Diethylenetriaminepentaacetic acid, 2,2',2'',2'''-((((Carboxymethyl)azanediyl)bis(ethane-2,1-diyl))bis(azanetriyl))tetraacetic acid, Diethylenetriamine-N,N,N',N'',N''-pentaacetic acid

D6518-5G D6518-50G D6518-10G D6518-100G
220122-585EA 91.94 USD
220122-585 220122-584 220122-583 220122-174
Diethylenetriaminepentaacetic acid ≥99%
Diethylenetriaminepentaacetic acid
Formula: C₁₄H₂₃N₃O₁₀
MW: 393.35 g/mol
Melting Pt: 220…222 °C
Density: 1.558 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004289
CAS Number: 67-43-6
EINECS: 200-652-8
UN: 3077
ADR: 9,III
Merck Index: 13,07202

Order Now

Specification Test Results

Assay 99% (titration)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR