5,7-Dichloro-8-quinolinol ≥99%

Synonyms: Chloroxine, 5,7-Dichloro-8-hydroxyquinoline

D64600-100G D64600-5G
220122-578EA 117.38 USD
220122-578 220122-579
5,7-Dichloro-8-quinolinol ≥99%
5,7-Dichloro-8-quinolinol
Formula: C₉H₅Cl₂NO
MW: 214.05 g/mol
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 183 °C
MDL Number: MFCD00006786
CAS Number: 773-76-2
EINECS: 212-258-3
Merck Index: 13,02193

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR