2,3-Dichloro-5,6-dicyano-p-benzoquinone

Synonyms: DDQ, 2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 2,3-Dichloro-5,6-dicyanoquinone, 4,5-Dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile

D60400-100G D60400-10G D60400-5G
220122-448EA 164.77 USD
220122-448 220122-449 220122-450
2,3-Dichloro-5,6-dicyano-p-benzoquinone
2,3-Dichloro-5,6-dicyano-p-benzoquinone
Formula: C₈Cl₂N₂O₂
MW: 227.01 g/mol
Boiling Pt: 396 °C (1013 hPa)
Melting Pt: 209…214 °C
Density: 1.713 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001593
CAS Number: 84-58-2
EINECS: 201-542-2
UN: 3439
ADR: 6.1,III
Merck Index: 13,03088

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR