1,4-Dichloro-2-butyne ≥99%

Warning

Synonyms: 1,4-Dichlorobut-2-yne

D59607-100G D59607-25G D59607-5G
220122-428EA 938 USD
220122-428 220122-429 220122-430
1,4-Dichloro-2-butyne ≥99%
1,4-Dichloro-2-butyne
Formula: C₄H₄Cl₂
MW: 122.98 g/mol
Boiling Pt: 165 °C (1013 hPa)
Melting Pt: –34.5 °C
Density: 1.2613 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000981
CAS Number: 821-10-3
EINECS: 212-474-8
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR