1,4-Dichlorobutane ≥97%

Danger

D59100-250G D59100-1KG
220121-528EA 59.88 USD
220121-528 220121-527
1,4-Dichlorobutane ≥97%
1,4-Dichlorobutane
Formula: ClH₂C(CH₂)₂CH₂Cl
MW: 127.01 g/mol
Boiling Pt: 153.9 °C (1013 hPa)
Melting Pt: –38 °C
Density: 1.1457 g/cm³ (20 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00001011
CAS Number: 110-56-5
EINECS: 203-778-1
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR