1,4-Dichlorobenzene ≥99%

Warning

Synonyms: p-Dichlorobenzene

D56829-100G D56829-3KG
220122-152EA 28.65 USD
220122-152 220122-153
1,4-Dichlorobenzene ≥99%
1,4-Dichlorobenzene
Formula: C₆H₄Cl₂
MW: 147 g/mol
Boiling Pt: 173.4 °C (1013 hPa)
Melting Pt: 48…50 °C
Density: 1.458 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00000604
CAS Number: 106-46-7
EINECS: 203-400-5
UN: 3077
ADR: 9,III
Merck Index: 13,03082

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR