2,6-Dichlorobenzaldehyde ≥99%

Warning

D56500-100G D56500-25G
220122-350EA 54.17 USD
220122-350 220122-351
2,6-Dichlorobenzaldehyde ≥99%
2,6-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 239 °C (1013 hPa)
Melting Pt: 69…70 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003307
CAS Number: 83-38-5
EINECS: 201-472-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR