1,8-Dichloro-9,10-anthraquinone

Synonyms: 1,8-Dichloroanthraquinone, 1,8-Dichloroanthracene-9,10-dione

D56403-25G D56403-100G
220122-345EA 64.62 USD
220122-345 220122-344
1,8-Dichloro-9,10-anthraquinone
1,8-Dichloro-9,10-anthraquinone
Formula: C₁₄H₆Cl₂O₂
MW: 277.11 g/mol
MDL Number: MFCD00001191
CAS Number: 82-43-9
EINECS: 201-420-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR