N,N-Dimethyl-1,4-phenylendiamine hydrochloride

Synonyms: N1,N1-Dimethylbenzene-1,4-diamine hydrochloride, N,N-Dimethyl-p-phenylenediamine monohydrochloride

D5004-100G D5004-10G D5004-25G
220122-230EA 53.88 USD
220122-230 220122-231 220122-232
N,N-Dimethyl-1,4-phenylendiamine hydrochloride
N,N-Dimethyl-1,4-phenylendiamine hydrochloride
Formula: C₈H₁₃ClN₂
MW: 172.66 g/mol
Melting Pt: 215 °C
MDL Number: MFCD00012990
CAS Number: 2052-46-2
EINECS: 218-146-0
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR