Dibutyl maleate ≥97%

Synonyms: Maleic acid dibutyl ester

D47102-250ML D47102-1L
220122-129EA 37.58 USD
220122-129 220122-128
Dibutyl maleate ≥97%
Dibutyl maleate
Formula: C₁₂H₂₀O₄
MW: 228.29 g/mol
Boiling Pt: 280 °C (1013 hPa)
Density: 0.9938 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009447
CAS Number: 105-76-0
EINECS: 203-328-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR