α,α'-Dibromo-p-xylene ≥97%

Warning

Synonyms: p-Xylylene dibromide, 1,4-Bis(bromomethyl)benzene

D44804-5G D44804-25G D44804-100G
220121-345EA 58.15 USD
220121-345 220121-344 220121-343
α,α'-Dibromo-p-xylene ≥97%
α,α'-Dibromo-p-xylene
Formula: C₈H₈Br₂
MW: 263.96 g/mol
Boiling Pt: 245 °C (lit.)
Melting Pt: 143…145 °C (lit.)
Density: 1.432 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000182
CAS Number: 623-24-5
EINECS: 210-781-1
UN: 3448
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR