α,α'-Dibromo-o-xylene ≥97%

Warning

Synonyms: 1,2-Bis(bromomethyl)benzene, o-Xylylene bromide, o-Xylylene dibromide, alpha,alpha'-Dibromo-o-xylene, o-Xylene dibromide

D44405-100G D44405-25G
220122-043EA 183.08 USD
220122-043 220122-044
α,α'-Dibromo-o-xylene ≥97%
α,α'-Dibromo-o-xylene
Formula: C₈H₈Br₂
MW: 263.96 g/mol
Boiling Pt: 140 °C (20 mmHg)
Melting Pt: 90…93 °C
Density: 1.96 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000175
CAS Number: 91-13-4
EINECS: 202-042-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR