3,3-Dimethylglutaric acid

Synonyms: 3,3-Dimethylpentanedioic acid, 3,3-Dimethyl-1,5-pentanedioic acid

D4379-25G D4379-250G D4379-100G
220122-026EA 65.05 USD
220122-026 220122-025 220122-024
3,3-Dimethylglutaric acid
3,3-Dimethylglutaric acid
Formula: C₇H₁₂O₄
MW: 160.17 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 98 - 101 °C
Density: 1.4278 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002716
CAS Number: 4839-46-7
EINECS: 225-425-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR