2,6-Dibromopyridine

D43115-25G D43115-100G
220122-018EA 121.69 USD
220122-018 220122-017
2,6-Dibromopyridine
2,6-Dibromopyridine
Formula: C₅H₃Br₂N
MW: 236.89 g/mol
Boiling Pt: 255 °C (1013 hPa)
Melting Pt: 116 - 119 °C
Storage Temperature: Ambient
MDL Number: MFCD00006223
CAS Number: 626-05-1
EINECS: 210-926-9
UN: 2811
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR