1,4-Dibromo-2-nitrobenzene ≥99%

Warning

Synonyms: 2,5-Dibromonitrobenzene

D42003-100G D42003-25G
220121-988EA 132.46 USD
220121-988 220121-989
1,4-Dibromo-2-nitrobenzene ≥99%
1,4-Dibromo-2-nitrobenzene
Formula: C₆H₃Br₂NO₂
MW: 280.9 g/mol
Melting Pt: 81…85 °C
Density: 2.374 g/cm³ (20 °C)
MDL Number: MFCD00007046
CAS Number: 3460-18-2
EINECS: 222-404-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR