N,N-Dimethyl-1,4-phenylenediammonium dichloride ≥99.00% (by titrimetric analysis) suitable for peroxidase analysis, Sigma-Aldrich®

Danger

Synonyms: N,N-Dimethyl-p-phenylenediamine dihydrochloride, N,N-Dimethyl-P-phenylenediamine 2HCl

M-Clarity™ quality level = MQ200

D4139-10G D4139-100G D4139-25G
220121-968EA 86.38 USD
220121-968 220121-967 220121-969
N,N-Dimethyl-1,4-phenylenediammonium dichloride ≥99.00% (by titrimetric analysis) suitable for peroxidase analysis, Sigma-Aldrich®
N,N-Dimethyl-1,4-phenylenediammonium dichloride
Formula: (CH₃)₂NC₆H₄NH₂·2HCl
MW: 209.12 g/mol
Melting Pt: 208 - 212 °C
Storage Temperature: Ambient
MDL Number: MFCD00012991
CAS Number: 536-46-9
EINECS: 208-635-7
UN: 2811
ADR: 6.1,II
Merck Index: 13,03280
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f64621224792e0a4614d3b6a46f0a95e29b130a8&vid=f012289c4ede1e0a60734047c7b7b455f6eb867e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (method) 50 mg/ml in water
Infrared spectrum Conforms to structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR