Barium diphenylamine-4-sulfonate

Warning

Synonyms: Diphenylamine-4-sulphonic acid barium salt, N-Phenylsulphanilic acid barium salt

D4130-25G D4130-5G D4130-50G
220121-962EA 473.85 USD
220121-962 220121-964 220121-963
Barium diphenylamine-4-sulfonate
Barium diphenylamine-4-sulphonate
Formula: C₂₄H₂₀BaN₂O₆S₂
MW: 633.89 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007497
CAS Number: 6211-24-1
EINECS: 228-278-0
UN: 1564
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36bef5e3b88ef2c1a87e7413bc720a14a5268931&vid=252019298b7070cfc5d7dd769eb30c8c67d6aae3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR