N,N-Dimethyl-1-naphthylamine

Warning

Synonyms: Dimethyl(1-naphthyl)amine, N,N-Dimethylnaphthalen-1-amine

D4011-10ML D4011-500ML D4011-100ML D4011-25ML
220121-940EA 95.48 USD
220121-940 220121-942 220121-939 220121-941
N,N-Dimethyl-1-naphthylamine
N,N-Dimethyl-1-naphthylamine

N,N-Dimethyl-1-naphthylamine is an aromatic amine and a dye.


  • N,N-Dimethyl-1-naphthylamine (1-Dimethylaminonaphthalene) is used to study nitrate reduction by organisms
  • Reduction of nitrate results in the formation of nitrite, which reacts with sulfanilic acid and 1-dimethylaminonaphthalene to form a red diazo dye

Formula: C₁₂H₁₃N
MW: 171.24 g/mol
Boiling Pt: 275 °C (1013 hPa)
Density: 1.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003919
CAS Number: 86-56-6
EINECS: 201-682-4
UN: 3082
ADR: 9,III
Merck Index: 12,03301
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a30cbb24dd2824343b2ff06291803d4293e4a81&vid=35f774c51db46f50ae94d7aaf07f4fec33ddfb16&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR