trans-1,4-Dibromo-2-butene ≥99%

Danger

Synonyms: (E)-1,4-Dibromobut-2-ene, (E)-1,4-Dibromo-2-butene

D39207-100G D39207-25G
220121-913EA 129.23 USD
220121-913 220121-914
trans-1,4-Dibromo-2-butene ≥99%
trans-1,4-Dibromo-2-butene
Formula: C₄H₆Br₂
MW: 213.9 g/mol
Boiling Pt: 203 °C (1013 hPa)
Melting Pt: 50…53 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000249
CAS Number: 821-06-7
EINECS: 212-472-7
UN: 2923
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR