(±)-1,4-Dibromo-2-butanol ≥85%, Technical Grade

Synonyms: 1,4-Dibromo-2-butanol

D39185-5G D39185-25G
220121-911EA 43.29 USD
220121-911 220121-910
(±)-1,4-Dibromo-2-butanol ≥85%, Technical Grade
(±)-1,4-Dibromo-2-butanol
Formula: BrH₂CCH(OH)CH₂CH₂Br
MW: 231.91 g/mol
Boiling Pt: 114 °C (1013 hPa)
Density: 2.023 g/cm³ (0 °C)
MDL Number: MFCD00000252
CAS Number: 19398-47-1
EINECS: 243-029-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR