2,4-Dibromoaniline ≥98%

D38405-5G D38405-25G D38405-100G
220121-881EA 75.28 USD
220121-881 220121-880 220121-879
2,4-Dibromoaniline ≥98%
2,4-Dibromoaniline
Formula: C₆H₅Br₂N
MW: 250.92 g/mol
Boiling Pt: 156 °C (21 mmHg)
Melting Pt: 80…82 °C
Density: 2.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007633
CAS Number: 615-57-6
EINECS: 210-434-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR