ω,4-Dibromoacetophenone ≥98%

Danger

Synonyms: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide

D38308-100G D38308-10G D38308-50G
220121-876EA 92.08 USD
220121-876 220121-877 220121-878
ω,4-Dibromoacetophenone ≥98%
ω,4-Dibromoacetophenone
Formula: C₈H₆Br₂O
MW: 277.94 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 107…108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000200
CAS Number: 99-73-0
EINECS: 202-783-6
UN: 3261
ADR: 8,II
Merck Index: 13,01412

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR