trans-1,4-Diphenyl-2-butene-1,4-dione ≥96%

Synonyms: 1,2-Dibenzoylethylene, trans-1,2-Dibenzoylethylene

D33209-25G D33209-5G
220120-281EA 316.62 USD
220120-281 220121-334
trans-1,4-Diphenyl-2-butene-1,4-dione ≥96%
trans-1,4-Diphenyl-2-butene-1,4-dione
Formula: C₁₆H₁₂O₂
MW: 236.27 g/mol
MDL Number: MFCD00003083
CAS Number: 959-28-4
EINECS: 213-498-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR