Dibenzothiophene

Synonyms: Dibenzo[b,d]thiophene

D32202-100G D32202-250G D32202-25G
220121-372EA 235.85 USD
220121-372 220121-373 220121-374
Dibenzothiophene
Dibenzothiophene
Formula: C₁₂H₈S
MW: 184.26 g/mol
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 95 - 98 °C
Density: 1.1295 g/cm³ (125 °C)
Flash Pt: 170 °C
MDL Number: MFCD00004969
CAS Number: 132-65-0
EINECS: 205-072-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR