1,3-Dimethyluric acid

Synonyms: 7,9-Dihydro-1,3-dimethyl-1H-purine-2,6,8(3H)-trione, 1,3-Dimethyl-1H-purine-2,6,8(3H,7H,9H)-trione

D2889-250MG D2889-100MG
220120-390EA 142.46 USD
220120-390 220121-825
1,3-Dimethyluric acid
1,3-Dimethyluric acid
Formula: C₇H₈N₄O₃
MW: 196.17 g/mol
Melting Pt: 412…415 °C
Density: 1.525 g/cm³ (25 °C)
MDL Number: MFCD00009740
CAS Number: 944-73-0
EINECS: 213-410-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR