1,4-Diazabicyclo[2.2.2]octane

Warning

Synonyms: DABCO

D27802-2KG D27802-25G D27802-500G D27802-100G
220121-809EA 534.62 USD
220121-809 220121-808 220121-810 220121-807
1,4-Diazabicyclo[2.2.2]octane
1,4-Diazabicyclo[2.2.2]octane
Formula: C₆H₁₂N₂
MW: 112.17 g/mol
Boiling Pt: 174 °C (1013 hPa)
Melting Pt: 155…158 °C
Density: 1.14 g/cm³ (20 °C)
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00006689
CAS Number: 280-57-9
EINECS: 205-999-9
UN: 1325
ADR: 4.1,III
Merck Index: 12,09801

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR