3,4-Diaminotoluene ≥97%

Synonyms: 1,2-Diamino-4-methylbenzene, Toluene-3,4-diamine

D26024-100G D26024-5G
220121-003EA 56.65 USD
220121-003 220121-004
3,4-Diaminotoluene ≥97%
3,4-Diaminotoluene
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00007728
CAS Number: 496-72-0
EINECS: 207-826-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR