Deoxycholic acid

Synonyms: 12α-Hydroxylithocholic acid, (R)-4-((3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-Dihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid, 3α,12α-Dihydroxy-5β-cholanic acid, 3α,12α-Dihydroxy-5β-cholan-24-oic acid, DCA, 12alpha-Hydroxylithocholic acid, 3α,12α-Dihydroxycholan-24-oic acid, 3α,12α-Dihydroxycholanic acid

D2510-100G D2510-500G D2510-10G
220121-171EA 163.6 USD
220121-171 220121-173 220121-172
Deoxycholic acid
Deoxycholic acid
Formula: C₂₄H₄₀O₄
MW: 392.58 g/mol
Boiling Pt: 504 °C (1013 hPa)
Melting Pt: 190 °C
MDL Number: MFCD00003673
CAS Number: 83-44-3
EINECS: 201-478-5
Merck Index: 13,02916

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR