1,3-Diaminopropane dihydrochloride ≥98%

Synonyms: Propane-1,3-diamine dihydrochloride, 1,3-Propanediamine dihydrochloride, Trimethylenediamine dihydrochloride

D23807-100G D23807-5G
220121-711EA 450.15 USD
220121-711 220121-712
1,3-Diaminopropane dihydrochloride ≥98%
1,3-Diaminopropane dihydrochloride
Formula: C₃H₁₂Cl₂N₂
MW: 147.05 g/mol
MDL Number: MFCD00012525
CAS Number: 10517-44-9
EINECS: 234-051-7

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR