1,3-Diaminopropane ≥99%

Synonyms: 1,3-Propanediamine, Trimethylenediamine

D23602-3KG D23602-500G D23602-25G
220121-707EA 325.23 USD
220121-707 220121-708 220121-706
1,3-Diaminopropane ≥99%
1,3-Diaminopropane
Formula: C₃H₁₀N₂
MW: 74.13 g/mol
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: –12 °C
Density: 0.863 g/cm³ (20 °C)
Flash Pt: 48 °C
Storage Temperature: Ambient
MDL Number: MFCD00008228
CAS Number: 109-76-2
EINECS: 203-702-7
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR