Dodecylamine ≥98%

Danger

Synonyms: 1-Dodecylamine, Dodecan-1-amine, 1-Aminododecane

D222208-1L D222208-100ML D222208-500ML
220121-681EA 130.31 USD
220121-681 220121-680 220121-682
Dodecylamine ≥98%
Dodecylamine
Formula: H₃C(CH₂)₁₀CH₂NH₂
MW: 185.35 g/mol
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 25…28 °C
Density: 0.8 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00008154
CAS Number: 124-22-1
EINECS: 204-690-6
UN: 3259
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR