1,12-Dodecanediol ≥99%

Synonyms: 1,12-Dihydroxy dodecane

D221309-100G D221309-25G
220121-610EA 140 USD
220121-610 220121-611
1,12-Dodecanediol ≥99%
1,12-Dodecanediol
Formula: C₁₂H₂₆O₂
MW: 202.34 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 78…79 °C
Density: 1.08 g/cm³ (20 °C)
MDL Number: MFCD00004755
CAS Number: 5675-51-4
EINECS: 227-133-9

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR