1,8-Diaminonaphthalene

Warning

Synonyms: 1,8-naphthylenediamine, 1,8-Naphthalenediamine, Naphthalene-1,8-diamine

D21405-5G D21405-100G
220121-590EA 38.88 USD
220121-590 220121-589
1,8-Diaminonaphthalene
1,8-Diaminonaphthalene
Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 64…65 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD00004033
CAS Number: 479-27-6
EINECS: 207-529-8
UN: 3259
ADR: 8,III
Merck Index: 12,06459

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR