1,5-Diaminonaphthalene

Warning

Synonyms: 1,5-naphthylenediamine, 1,5-Naphthalenediamine

D21200-25G D21200-100G
220121-579EA 78.62 USD
220121-579 220121-578
1,5-Diaminonaphthalene
1,5-Diaminonaphthalene
Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 381 °C (1013 hPa)
Melting Pt: 188 °C
Density: 1.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004029
CAS Number: 2243-62-1
EINECS: 218-817-8
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR