Diphenylmethane ≥99%

Warning

D209317-1KG D209317-25G D209317-500G
220121-566EA 142.15 USD
220121-566 220121-567 220121-568
Diphenylmethane ≥99%
Diphenylmethane
Formula: C₁₃H₁₂
MW: 168.24 g/mol
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 22…25 °C
Density: 0.9985 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004781
CAS Number: 101-81-5
EINECS: 202-978-6
UN: 3077
ADR: 9,III
Merck Index: 13,03360

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR