trans,trans-1,4-Diphenyl-1,3-butadiene

Synonyms: (1E,3E)-1,4-Diphenylbuta-1,3-diene

D206008-25G D206008-5G
220121-498EA 156.15 USD
220121-498 220120-381
trans,trans-1,4-Diphenyl-1,3-butadiene
trans,trans-1,4-Diphenyl-1,3-butadiene
Formula: C₁₆H₁₄
MW: 206.29 g/mol
MDL Number: MFCD00004791
CAS Number: 538-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR